6-Bromo-2(3H)-benzoxazolone ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 6-Bromo-2-benzoxazolinone, 6-Bromo-2-benzoxazolone, 6-Bromo-2-hydroxybenzoxazole, 6-Bromobenzo[d]oxazol-2(3H)-one

B5290-1G B5290-5G
TCB5290-1GEA 98.88 USD
TCB5290-1G TCB5290-5G
6-Bromo-2(3H)-benzoxazolone ≥98.0% (by GC)
6-Bromo-2(3H)-benzoxazolone
Formula: C₇H₄BrNO₂
MDL Number: MFCD00694727
CAS Number: 19932-85-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=291d30781fbd59d10c83a1193fa9b0d288d0d760&vid=ec54952a4edc3fcf6d779060093fe18b23f82751&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 193.0 to 197.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR