4-(n-Butoxy)benzenesulfonyl chloride ≥98.0% (by GC)

Supplier: TCI America
Danger

Synonyms: 4-Butoxybenzenesulphonyl chloride, 4-Butoxybenzene-1-sulphonyl chloride, 4-Butoxyphenylsulphonyl chloride

B2587-1G B2587-5G
TCB2587-1GEA 42.23 USD
TCB2587-1G TCB2587-5G
4-(n-Butoxy)benzenesulfonyl chloride ≥98.0% (by GC)
4-(n-Butoxy)benzenesulfonyl chloride
Formula: C₁₀H₁₃ClO₃S
MW: 248.73 g/mol
Boiling Pt: 121 °C
Melting Pt: 19 °C
MDL Number: MFCD00052344
CAS Number: 1138-56-3
UN: 3265
ADR: 8,II

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR