4-Aminopyrimidine-5-carbonitrile ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

A1214-1G A1214-5G
TCA1214-1GEA 22.66 USD
TCA1214-1G TCA1214-5G
4-Aminopyrimidine-5-carbonitrile ≥98.0% (by GC, titration analysis)
4-Aminopyrimidine-5-carbonitrile
Formula: C₅H₄N₄
MW: 120.11 g/mol
MDL Number: MFCD00054552
CAS Number: 16357-69-0
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=611999e2be17ed6f8b23e1ecaeab2f2c5e11d36d&vid=8228e0b634c9d1d80879ae01e589ea6f72baccc8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR