Zofenopril Calcium ≥97.0% (by HPLC)

Supplier: TCI America

Z0041-100MG Z0041-25MG
TCZ0041-100MGEA 97.85 USD
TCZ0041-100MG TCZ0041-25MG
Zofenopril Calcium ≥97.0% (by HPLC)
Zofenopril Calcium
Formula: C₄₄H₄₄CaN₂O₈S₄
Melting Pt: 246 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08704648
CAS Number: 81938-43-4
Merck Index: 14,10186
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=82f6fb2c6ace7299731fa65864f7e1bfc1fa522c&vid=8b1095084e018480a40197aa35272b628b74852d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 97.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR