WAY-100635 Maleate ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: WAY-100635 maleate, N-[2-[4-(2-methoxyphenyl)-1-piperazinyl]ethyl]-N-2-pyridinyl-cyclohexanecarboxamide 2Z-butenedioate, N-[2-[4-(2-Methoxyphenyl)piperazin-1-yl]ethyl]-N-(pyridin-2-yl)cyclohexanecarboxamide maleate

W0016-10MG W0016-50MG
TCW0016-10MGEA 186.43 USD
TCW0016-10MG TCW0016-50MG
WAY-100635 Maleate ≥98.0% (by HPLC)
WAY-100635 Maleate
Formula: C₂₅H₃₄N₄O₂·C₄H₄O₄
MW: 538.65 g/mol
MDL Number: MFCD01321056
CAS Number: 1092679-51-0

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR