Poly(2-ethyl-2-oxazoline) Azide Terminated n=∼50, ULTROXA®
Supplier: TCI America
Synonyms:
alpha-Methyl-omega-azido-poly[[(1-oxopropyl)imino]-1,2-ethanediyl]
U0132-100MG
U0132-500MG
TCU0132-100MGEA
91.67
USD
TCU0132-100MG
TCU0132-500MG
Poly(2-ethyl-2-oxazoline) Azide Terminated n=∼50, ULTROXA®
Poly(2-ethyl-2-oxazoline) Azide Terminated
Storage Temperature:
Refrigerator |
CAS Number:
1431850-88-2 |
Specification Test Results
Appearance | White to Almost white powder to lump |
Average molecular weight | 4,000 to 6,000 |
Molecular weight distribution | max. 1.15 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...