(Triethoxysilyl)methyl methacrylate ≥97.0% (by GC) stabilised

Supplier: TCI America
Warning

Synonyms: Methacrylic acid (Triethoxysilyl)methyl ester

Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

T3852-25G T3852-5G
TCT3852-25GEA 255.44 USD
TCT3852-25G TCT3852-5G
(Triethoxysilyl)methyl methacrylate ≥97.0% (by GC) stabilised
(Triethoxysilyl)methyl methacrylate
Formula: C₁₁H₂₂O₅Si
MW: 262.38 g/mol
Boiling Pt: 97 °C
MDL Number: MFCD00156470
CAS Number: 5577-72-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c120243744e787e44669247a8a822c88d37fa0cd&vid=5416cbb05cc55586e6c3ea7441d8191c0bff6bbd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR