4-(Trifluoromethyl)benzylamine hydrobromide ≥98.0% (by HPLC)
Supplier: TCI America
|
Warning
|
Synonyms:
[4-(Trifluoromethyl)phenyl]methanamine hydrobromide, 4-(Trifluoromethyl)benzylammonium bromide, 4-Aminomethylbenzotrifluoride hydrobromide
T3837-1G
T3837-5G
TCT3837-1GEA
31.93
USD
TCT3837-1G
TCT3837-5G
4-(Trifluoromethyl)benzylamine hydrobromide ≥98.0% (by HPLC)
4-(Trifluoromethyl)benzylamine hydrobromide
|
Specification Test Results
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Nonaqueous Titration) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...