1H-Pyrazole-3-boronic acid pinacol ester ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole, 2-(1H-Pyrazol-3-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 1H-Pyrazole-3-boronic acid Pinacol ester

T3829-1G T3829-5G
TCT3829-1GEA 65.92 USD
TCT3829-1G TCT3829-5G
1H-Pyrazole-3-boronic acid pinacol ester ≥98.0% (by titrimetric analysis)
1H-Pyrazole-3-boronic acid pinacol ester
Formula: C₉H₁₅BN₂O₂
MW: 194.04 g/mol
MDL Number: MFCD07368044
CAS Number: 844501-71-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8f45bbc1c304c31e916f7eb066f9cbdce94c9cba&vid=ac21e54fea6eb717cb20794ee26759a0ea8de1c5&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralisation titration) min. 98.0 %
Melting point 105.0 to 109.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR