Tirapazamine ≥98.0% (by HPLC)

Supplier: TCI America
Warning

T3823-250MG T3823-50MG
TCT3823-250MGEA 684.95 USD
TCT3823-250MG TCT3823-50MG
Tirapazamine ≥98.0% (by HPLC)
Tirapazamine
Formula: C₇H₆N₄O₂
Melting Pt: 230 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00132954
CAS Number: 27314-97-2
Merck Index: 14,09461
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fad4172d036da0ae5b38088e9fd2d98e743cee45&vid=52fb0606002bf5a86161d6b2ba141f532579ce6e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Orange to Red powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 30.00 to 33.00 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR