Thieno[2,3-d]pyrimidin-4(3H)-one ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: Thieno[2,3-d]pyrimidin-4(1H)-one, 3,4-Dihydro-4-oxothieno[2,3-d]pyrimidine

T3792-1G T3792-5G
TCT3792-1GEA 36.05 USD
TCT3792-1G TCT3792-5G
Thieno[2,3-d]pyrimidin-4(3H)-one ≥98.0% (by HPLC, titration analysis)
Thieno[2,3-d]pyrimidin-4(3H)-one
Formula: C₆H₄N₂OS
MW: 152.1738 g/mol
Melting Pt: 245 °C
MDL Number: MFCD00706456
CAS Number: 14080-50-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=43e6125deeef25f04c3c87d0fe3fb627d373aa5a&vid=f926b653594ec82a99bd076061c1b10b7cd22628&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR