2,4,4-Trimethylpentan-2-amine hydroiodide ≥97.0% (by titrimetric analysis)

Supplier: TCI America
Warning

Synonyms: 1,1,3,3-Tetramethylbutylamine Hydroiodide, tert-Octylamine Hydroiodide, tert-Octylammonium iodide

T3785-1G T3785-5G
TCT3785-1GEA 35.02 USD
TCT3785-1G TCT3785-5G
2,4,4-Trimethylpentan-2-amine hydroiodide ≥97.0% (by titrimetric analysis)
2,4,4-Trimethylpentan-2-amine hydroiodide
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=99e852f0c81c99b52d326427ee213a2a42be6b59&vid=38abfdb8a3924ab5b0ae5ad4737141d4e82c9563&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR