1-(Trifluoromethyl)cyclopropanecarboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: 1-(Trifluoromethyl)cyclopropane-1-carboxylic acid

T3765-1G T3765-200MG
TCT3765-1GEA 57.68 USD
TCT3765-1G TCT3765-200MG
1-(Trifluoromethyl)cyclopropanecarboxylic acid ≥98.0% (by GC, titration analysis)
1-(Trifluoromethyl)cyclopropanecarboxylic acid
Formula: C₅H₅F₃O₂
MW: 154.09 g/mol
Boiling Pt: 184 °C
MDL Number: MFCD03093070
CAS Number: 277756-46-4
UN: 3261
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=58b85c3ed674f2741b3a405b3b8d74318b31a59f&vid=b42ff4f35a2e77d731c9600f15973329fa1fdb25&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralisation titration) min. 98.0 %
Melting point 86.0 to 90.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR