Tectorigenin ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 4',5,7-Trihydroxy-6-methoxyisoflavone, 5,7-Dihydroxy-3-(4-hydroxyphenyl)-6-methoxychromone

T3729-10MG T3729-50MG
TCT3729-10MGEA 142.14 USD
TCT3729-10MG TCT3729-50MG
Tectorigenin ≥98.0% (by HPLC)
Tectorigenin
Formula: C₁₆H₁₂O₆
MW: 300.27 g/mol
MDL Number: MFCD00597094
CAS Number: 548-77-6
Merck Index: 14,09107
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0b7b48cbed93abc0989162032338ce56dc475f62&vid=f2d7819b03ae15d83efbff8e60bc5da3527c30d0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Carbon) 62.10 to 64.60 %
Melting point 228.0 to 232.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR