2,3,5,6-Tetrachloro-4-(methylsulfonyl)pyridine ≥95.0% (by GC)

Supplier: TCI America
Warning

T3670-1G T3670-5G
TCT3670-1GEA 32.96 USD
TCT3670-1G TCT3670-5G
2,3,5,6-Tetrachloro-4-(methylsulfonyl)pyridine ≥95.0% (by GC)
2,3,5,6-Tetrachloro-4-(methylsulfonyl)pyridine
Formula: C₆H₃Cl₄NO₂S
MDL Number: MFCD00054553
CAS Number: 13108-52-6
EINECS: 236-035-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=eddeb5b602ac3f8c14fb7ccd5e4e35102824b33f&vid=8028bcfe627bb15e0dde5b401235a961c63e1ad2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 95.0 %
Melting point 146.0 to 151.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR