Thifluzamide ≥98.0% (by GC)

Supplier: TCI America
Warning

T3648-1G T3648-250MG
TCT3648-1GEA 404.79 USD
TCT3648-1G TCT3648-250MG
Thifluzamide ≥98.0% (by GC)
Thifluzamide
Formula: C₁₃H₆Br₂F₆N₂O₂S
MDL Number: MFCD03428071
CAS Number: 130000-40-7
UN: 3077
ADR: 9,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e1a2a592ee3898a729948f43098ed7f857cfa136&vid=a02b4606b3764fc0229e78c3177b0a79e23a6836&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Gray to Red powder to crystal
Purity (GC) min. 98.0 %
Melting point 175.0 to 179.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR