3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-9-(p-tolyl)-9H-carbazole ≥98.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: 9-(p-Tolyl)-9H-carbazole-3-boronic acid Pinacol ester, 2-[9-(p-Tolyl)-9H-carbazol-3-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

T3612-1G T3612-200MG
TCT3612-1GEA 199.82 USD
TCT3612-1G TCT3612-200MG
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-9-(p-tolyl)-9H-carbazole ≥98.0% (by HPLC)
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-9-(p-tolyl)-9H-carbazole
Formula: C₂₅H₂₆BNO₂
MW: 383.30 g/mol
CAS Number: 1345614-94-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3e4cc57496fd4df33390f737851cca1e7e397141&vid=f2f3d43032b038e56587a5d9d2aaf00bdad2bd28&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralisation titration) min. 97.0 %
Melting point 164.0 to 168.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR