RG-13022 ≥98.0% (by GC)

Supplier: TCI America

T3575-100MG T3575-25MG
TCT3575-100MGEA 539.72 USD
TCT3575-100MG TCT3575-25MG
RG-13022 ≥98.0% (by GC)
RG-13022
Formula: C₁₆H₁₄N₂O₂
Storage Temperature: Refrigerator
MDL Number: MFCD00236444
CAS Number: 136831-48-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ae41b62f37e952da7213404e86ad1ae08f39ae8&vid=9a3f55a5632097f1393565c538bb4307a059a47e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 116.0 to 120.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR