Triallyl(methyl)silane ≥95.0% (by GC)
Supplier: TCI America
|
Warning
|
T3549-1ML
T3549-5ML
TCT3549-1MLEA
87.55
USD
TCT3549-1ML
TCT3549-5ML
Triallyl(methyl)silane ≥95.0% (by GC)
Triallyl(methyl)silane
Formula:
C₁₀H₁₈Si Boiling Pt: 180 °C |
MDL Number:
MFCD00800604 CAS Number: 1112-91-0 UN: 1993 ADR: 3,III |
|
Specification Test Results
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...