3-(Trifluoromethoxy)benzenesulfonyl chloride ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: 3-(Trifluoromethoxy)benzene-1-sulphonyl chloride

T3508-1G T3508-5G
TCT3508-1GEA 30.9 USD
TCT3508-1G TCT3508-5G
3-(Trifluoromethoxy)benzenesulfonyl chloride ≥98.0% (by GC, titration analysis)
3-(Trifluoromethoxy)benzenesulphonyl chloride
Formula: C₇H₄ClF₃O₃S
MW: 260.62 g/mol
MDL Number: MFCD01091016
CAS Number: 220227-84-9
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c382c865e2c5f84c5bcaea896e13b6b55756f6b&vid=729782e2d87d5f7465dbba023428217642513a50&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR