trans,trans-3,4,5-Trifluoro-4'-(4'-pentylbicyclohexyl-4-yl)biphenyl ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 3,4,5-Trifluoro-4'-[(trans,trans)-4'-pentyl[1,1'-bicyclohexyl]-4-yl]-1,1'-biphenyl

T3482-1G T3482-5G
TCT3482-1GEA 51.5 USD
TCT3482-1G TCT3482-5G
trans,trans-3,4,5-Trifluoro-4'-(4'-pentylbicyclohexyl-4-yl)biphenyl ≥98.0% (by GC)
trans,trans-3,4,5-Trifluoro-4'-(4'-pentylbicyclohexyl-4-yl)biphenyl
Formula: C₂₉H₃₇F₃
MW: 442.61 g/mol
MDL Number: MFCD11053475
CAS Number: 137529-43-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=268013ae04bc4dbf744c953f922feee93be8ac16&vid=c666bc2771441e1d77a4ad578becbdacf11f515c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR