2,3,6-Trichloropyridine ≥97.0% (by GC)

Supplier: TCI America
Warning

T3428-25G T3428-5G
TCT3428-25GEA 204.97 USD
TCT3428-25G TCT3428-5G
2,3,6-Trichloropyridine ≥97.0% (by GC)
2,3,6-Trichloropyridine
Formula: C₅H₂Cl₃N
Boiling Pt: 234 °C
MDL Number: MFCD00463516
CAS Number: 6515-09-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5f2f4c715531a9a6586e7477d06387a53330b1df&vid=5bc5ee18520d2e2d525df8f8f33530f1861c3084&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 97.0 %
Melting point 64.0 to 68.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR