N-Methoxy-N-methyltrifluoroacetamide ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: 2,2,2-Trifluoro-N-methoxy-N-methylacetamide

T3326-25G T3326-5G
TCT3326-25GEA 203.94 USD
TCT3326-25G TCT3326-5G
N-Methoxy-N-methyltrifluoroacetamide ≥98.0% (by GC)
N-Methoxy-N-methyltrifluoroacetamide
Formula: C₄H₆F₃NO₂
MW: 157.09 g/mol
Boiling Pt: 52 °C
Flash Pt: 47 °C
MDL Number: MFCD00077565
CAS Number: 104863-67-4
UN: 1993
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=203c52fa903e00aafc654554f279cbba06138789&vid=e09b1c2020c1f50b7236d980719a0a4a99ebf477&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR