4-(Trifluoromethyl)nicotinic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

T2742-1G T2742-5G
TCT2742-1GEA 46.35 USD
TCT2742-1G TCT2742-5G
4-(Trifluoromethyl)nicotinic acid ≥98.0% (by GC, titration analysis)
4-(Trifluoromethyl)nicotinic acid
Formula: C₇H₄F₃NO₂
MDL Number: MFCD00082626
CAS Number: 158063-66-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9d6d39740cb6d838fdca6afb67f03cfa40ec4b7c&vid=63d179ea0701beb2b3f7c8fadfb45dd392e57024&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralisation titration) min. 98.0 %
Melting point 137.0 to 141.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR