Dihydroxybis(ammonium lactato)titanium(IV) ∼40% in 2-propanol/water

Supplier: TCI America
Danger

T2611-25G T2611-500G
TCT2611-25GEA 29.87 USD
TCT2611-25G TCT2611-500G
Dihydroxybis(ammonium lactato)titanium(IV) ∼40% in 2-propanol/water
Dihydroxybis(ammonium lactato)titanium(IV)
Formula: C₆H₂₂N₂O₈Ti
MW: 298.115 g/mol
MDL Number: MFCD00072736
CAS Number: 65104-06-5
EINECS: 265-409-0
UN: 1219
ADR: 3 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=811a0957194295cc9af5185c34bdba19048ff6c9&vid=1a3224c17167fc49fd7f34f13cea8da2dbdab966&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Concentration (Gravimetric Method) 37.0 to 45.0 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR