(+/-)-Phenyl(pyridin-2-yl)methanol ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Phenyl(pyridin-2-yl)methanol, (+/-)-2-(alpha-Hydroxybenzyl)pyridine

P2755-1G P2755-5G
TCP2755-1GEA 136.99 USD
TCP2755-1G TCP2755-5G
(+/-)-Phenyl(pyridin-2-yl)methanol ≥98.0% (by GC)
(+/-)-Phenyl(pyridin-2-yl)methanol
Formula: C₁₂H₁₁NO
MW: 185.23 g/mol
MDL Number: MFCD00186514
CAS Number: 14159-57-0

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR