2-Phenyl-1,3-dithiolane ≥98.0% (by GC)

Supplier: TCI America

P2742-1G P2742-5G
TCP2742-1GEA 47.38 USD
TCP2742-1G TCP2742-5G
2-Phenyl-1,3-dithiolane ≥98.0% (by GC)
2-Phenyl-1,3-dithiolane
Formula: C₉H₁₀S₂
Boiling Pt: 166 °C
Melting Pt: 24 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01729637
CAS Number: 5616-55-7

Order Now

Specification Test Results

Appearance White to Almost white powder to lump
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR