Pentafluorophenyl Diphenylphosphinate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Diphenylphosphinic acid Pentafluorophenyl ester, FDPP, Perfluorophenyl diphenylphosphinate

P2726-1G P2726-5G
TCP2726-1GEA 36.05 USD
TCP2726-1G TCP2726-5G
Pentafluorophenyl Diphenylphosphinate ≥98.0% (by GC)
Pentafluorophenyl Diphenylphosphinate
Formula: C₁₈H₁₀F₅O₂P
MW: 384.24 g/mol
Boiling Pt: 153 °C
MDL Number: MFCD00192380
CAS Number: 138687-69-1

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 55.0 to 59.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR