1-[1-Phenyl-3,4-dihydro-1H-pyrido[3,4-b]indol-2(9H)-yl]pentan-1-one ≥98.0% (by HPLC)

Supplier: TCI America

Synonyms: 1-(1-Phenyl-1,3,4,9-tetrahydro-2H-pyrido[3,4-b]indol-2-yl)pentan-1-one

P2695-10MG P2695-50MG
TCP2695-10MGEA 172.01 USD
TCP2695-10MG TCP2695-50MG
1-[1-Phenyl-3,4-dihydro-1H-pyrido[3,4-b]indol-2(9H)-yl]pentan-1-one ≥98.0% (by HPLC)
1-[1-Phenyl-3,4-dihydro-1H-pyrido[3,4-b]indol-2(9H)-yl]pentan-1-one
Formula: C₂₂H₂₄N₂O
MW: 332.45 g/mol
MDL Number: MFCD01496905
CAS Number: 334939-35-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f315fa6c93cecef7b3724cdb3f0d688c6713a928&vid=0f13de209db9ed14c2aaf337ee7e9b394b034236&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Elemental analysis (Nitrogen) 8.00 to 8.90 %
Melting point 187.0 to 191.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR