4-(Pyridin-3-yl)pyrimidin-2-amine ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 4-(3-Pyridinyl)-2-pyrimidinamine

P2666-1G P2666-5G
TCP2666-1GEA 89.61 USD
TCP2666-1G TCP2666-5G
4-(Pyridin-3-yl)pyrimidin-2-amine ≥98.0% (by GC, titration analysis)
4-(Pyridin-3-yl)pyrimidin-2-amine
Formula: C₉H₈N₄
MW: 172.19 g/mol
MDL Number: MFCD01317832
CAS Number: 66521-66-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dafc7e2ad06e4288ea09c056c650f7cb8c61234a&vid=408177477a25626f28360f598692c2029eddb653&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 189.0 to 193.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR