1-Phenyl-3-(p-tolyl)-1H-pyrazole-4-carboxaldehyde ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 1-Phenyl-3-(p-tolyl)-1H-pyrazole-4-carbaldehyde

P2657-1G P2657-200MG
TCP2657-1GEA 96.82 USD
TCP2657-1G TCP2657-200MG
1-Phenyl-3-(p-tolyl)-1H-pyrazole-4-carboxaldehyde ≥98.0% (by GC)
1-Phenyl-3-(p-tolyl)-1H-pyrazole-4-carboxaldehyde
Formula: C₁₇H₁₄N₂O
MW: 262.31 g/mol
MDL Number: MFCD00744772
CAS Number: 36640-52-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2cc75d797b2eca5ddeb414300b8a008cd71141df&vid=40753f7bba1c69e3b0dbf9a296c611445d4c58d4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 117.0 to 121.0 deg-C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR