Potassium(4-chlorophenyl)trifluoroborate ≥98.0% (by HPLC)

Supplier: TCI America
Warning

Synonyms: (4-Chlorophenyl)trifluoroboric acid potassium salt, Potassium (4-chlorophenyl)trifluoroborate

P2654-25G P2654-5G
TCP2654-25GEA 290.46 USD
TCP2654-25G TCP2654-5G
Potassium(4-chlorophenyl)trifluoroborate ≥98.0% (by HPLC)
Potassium(4-chlorophenyl)trifluoroborate
Formula: C₆H₄BClF₃K
MW: 218.45 g/mol
MDL Number: MFCD04115770
CAS Number: 661465-44-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dd03527dacd301a29e9f023470c6a6ff4ac72bd6&vid=012b350a2bbf40af9e9c8a44fa4a7351a56ad662&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 97.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR