2-Phenylimidazo[1,2-a]pyridine ≥98.0% (by GC)

Supplier: TCI America

P2632-1G P2632-5G
TCP2632-1GEA 62.83 USD
TCP2632-1G TCP2632-5G
2-Phenylimidazo[1,2-a]pyridine ≥98.0% (by GC)
2-Phenylimidazo[1,2-a]pyridine
Formula: C₁₃H₁₀N₂
Boiling Pt: 245 °C
MDL Number: MFCD00219254
CAS Number: 4105-21-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5ccb6ca7fc57461261676edb6eadba915ef157ee&vid=a1036223dcd4f6a18eec1c737871f323e65a4a2a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 133.0 to 137.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR