Spautin-1 ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
6-Fluoro-N-(4-fluorobenzyl)quinazolin-4-amine, 6-Fluoro-N-[(4-fluorophenyl)methyl]-4-quinazolinamine
P2613-10MG
P2613-50MG
TCP2613-10MGEA
166.86
USD
TCP2613-10MG
TCP2613-50MG
Spautin-1 ≥98.0% (by GC, titration analysis)
Spautin-1
Formula:
C₁₅H₁₁F₂N₃ MW: 271.27 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD16962292 CAS Number: 1262888-28-7 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 209.0 to 213.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...