2-Phenoxyphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

Synonyms: 2-Phenoxybenzeneboronic acid

P2539-1G P2539-5G
TCP2539-1GEA 36.05 USD
TCP2539-1G TCP2539-5G
2-Phenoxyphenylboronic acid (contains varying amounts of Anhydride)
2-Phenoxyphenylboronic acid
Formula: C₁₂H₁₁BO₃
MW: 214.03 g/mol
Melting Pt: 114 °C
MDL Number: MFCD01001592
CAS Number: 108238-09-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1d05473dae39679b855984812fefba21c390b736&vid=43bc7c79f9410d1e67dec38f47036f203d22c854&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 98.0 to 109.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR