Potassium (Phthalimidomethyl)trifluoroborate ≥98.0% (by total nitrogen basis)

Supplier: TCI America
Warning

P2514-1G P2514-5G
TCP2514-1GEA 131.84 USD
TCP2514-1G TCP2514-5G
Potassium (Phthalimidomethyl)trifluoroborate ≥98.0% (by total nitrogen basis)
Potassium (Phthalimidomethyl)trifluoroborate
Formula: C₉H₆BF₃KNO₂
MW: 267.06 g/mol
Melting Pt: 276 °C
MDL Number: MFCD19000584
CAS Number: 1001671-72-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2c04f78a1c9f108cc8c7786a6f09632027c371d4&vid=cb59c01fda0c2bd159fe4a9c78415ee29c8c6c0d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (with Total Nitrogen) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR