4-(1-Pyrazolyl)benzonitrile ≥98.0% (by GC)

Supplier: TCI America
Warning

P2450-1G P2450-5G
TCP2450-1GEA 36.05 USD
TCP2450-1G TCP2450-5G
4-(1-Pyrazolyl)benzonitrile ≥98.0% (by GC)
4-(1H-Pyrazol-3-yl)benzonitrile
Formula: C₁₀H₇N₃
MDL Number: MFCD09033533
CAS Number: 25699-83-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e53626ab07efebb84561ddb27e13cf6ed3847eb6&vid=c61b0f9e7ebc6113fb25626e1ad1656d9f87ce64&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 87.0 to 91.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR