(Phenylhydrazono)malononitrile ≥96.0% (by GC)

Supplier: TCI America
Danger

P2346-25G P2346-5G
TCP2346-25GEA 167.89 USD
TCP2346-25G TCP2346-5G
(Phenylhydrazono)malononitrile ≥96.0% (by GC)
(Phenylhydrazono)malononitrile
Formula: C₉H₆N₄
MDL Number: MFCD00195983
CAS Number: 306-18-3
UN: 3439
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=45c19440c46f8c0723a695b919adff694a08b5e8&vid=9fa58543da1dbe800c9b5f7592a40871f0fb2ad0&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Light yellow to Yellow to Orange powder to crystal
Purity (GC) min. 96.0 %
Melting point 131.0 to 135.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR