2-Phthalimidoethanesulfonyl chloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

Synonyms: 2-(1,3-Dioxoisoindolin-2-yl)ethanesulphonylchloride

P2049-1G P2049-5G
TCP2049-1GEA 86.52 USD
TCP2049-1G TCP2049-5G
2-Phthalimidoethanesulfonyl chloride ≥98.0% (by titrimetric analysis)
2-Phthalimidoethanesulfonyl chloride
Formula: C₁₀H₈ClNO₄S
MW: 273.7 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD01861217
CAS Number: 4403-36-5
UN: 3261
ADR: 8 II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=05fbd58877f0573f3949141499d4810bed5c3165&vid=81d9ee611b5404d4c9eabea486f9235c2f27f69c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Argentmetric Titration) min. 98.0 %
Melting point 160.0 to 164.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR