Octan-2-yl (2,4-Dichlorophenoxy)acetate ≥96.0% (by GC)

Supplier: TCI America

Synonyms: 2,4-D Meptyl ester, 3-(2,4-Dichlorophenoxy)octan-2-yl acetate, 2,4-D IOE, 2,4-D Octan-2-yl ester, (2,4-Dichlorophenoxy)acetic acid Octan-2-yl ester

O0518-100MG O0518-1G
TCO0518-100MGEA 84.46 USD
TCO0518-100MG TCO0518-1G
Octan-2-yl (2,4-Dichlorophenoxy)acetate ≥96.0% (by GC)
Octan-2-yl (2,4-Dichlorophenoxy)acetate
Formula: C₁₆H₂₂Cl₂O₃
MW: 333.25 g/mol
CAS Number: 1917-97-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa7d337cb3962e9e9f36325f34f2727e22ec634b&vid=e6e51cbc242bace250eaa342f2cf669620164a95&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR