3,4-Dihydro-3-oxo-2H[1,4]benzothiazin-2ylacetic acid ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

O0504-1G O0504-5G
TCO0504-1GEA 99.91 USD
TCO0504-1G TCO0504-5G
3,4-Dihydro-3-oxo-2H[1,4]benzothiazin-2ylacetic acid ≥98.0% (by HPLC, titration analysis)
3,4-Dihydro-3-oxo-2H[1,4]benzothiazin-2ylacetic acid
Formula: C₁₀H₉NO₃S
MDL Number: MFCD00087524
CAS Number: 6270-74-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d3ea73215fcc39bc45b10e1f81e4000139dc0f6&vid=d58536b9f830196b0128cc57327e89c828bc84ac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralisation titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR