Oxazolo[4,5-b]pyridin-2(3H)-one ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2,3-Dihydropyrido[2,3-d]oxazol-2-one

O0408-25G O0408-5G
TCO0408-25GEA 189.52 USD
TCO0408-25G TCO0408-5G
Oxazolo[4,5-b]pyridin-2(3H)-one ≥98.0% (by GC, titration analysis)
Oxazolo[4,5-b]pyridin-2(3H)-one
Formula: C₆H₄N₂O₂
MDL Number: MFCD00204215
CAS Number: 60832-72-6

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralisation titration) min. 98.0 %
Melting point 212.0 to 216.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR