9-(1-Naphthyl)-10-(2-naphthyl)anthracene ≥98.0% (by HPLC)

Supplier: TCI America

N1135-1G N1135-5G
TCN1135-1GEA 92.7 USD
TCN1135-1G TCN1135-5G
9-(1-Naphthyl)-10-(2-naphthyl)anthracene ≥98.0% (by HPLC)
9-(1-Naphthyl)-10-(2-naphthyl)anthracene
Formula: C₃₄H₂₂
MDL Number: MFCD20040456
CAS Number: 855828-36-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5db580c32fb7e5a616ebb87467e9c83411cc982d&vid=f1c7cc69f51434131d49a1162f421fb578dd51cd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (HPLC) min. 98.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR