4-Mercapto-4-methyl-2-pentanone ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 4-Mercapto-4-methylpentan-2-one

M3271-25G M3271-5G
TCM3271-25GEA 244.11 USD
TCM3271-25G TCM3271-5G
4-Mercapto-4-methyl-2-pentanone ≥98.0% (by GC, titration analysis)
4-Mercapto-4-methyl-2-pentanone
Formula: C₆H₁₂OS
MW: 132.23 g/mol
Boiling Pt: 57 °C
Flash Pt: 54 °C
MDL Number: MFCD00085208
CAS Number: 19872-52-7
EINECS: 243-386-8
UN: 3336
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Iodometric Titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR