3-(4-Methyl-1-piperazinylmethyl)benzeneboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 4,4,5,5-Tetramethyl-2-[3-(4-Methyl-1-piperazinylmethyl)phenyl]-1,3,2-dioxaborolane, 1-Methyl-4-(3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl)piperazine, 3-(4-Methyl-1-piperazinylmethyl)benzeneboronic acid Pinacol ester, 1-Methyl-4-[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzyl]piperazine

M3245-5G M3245-1G
TCM3245-5GEA 237.93 USD
TCM3245-5G TCM3245-1G
3-(4-Methyl-1-piperazinylmethyl)benzeneboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
3-(4-Methyl-1-piperazinylmethyl)benzeneboronic acid pinacol ester
Formula: C₁₈H₂₉BN₂O₂
MW: 316.25 g/mol
MDL Number: MFCD09266181
CAS Number: 883738-27-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=74c5a59227bb95987ccc54aac831c9d61047d68e&vid=1dad57778f7f9c32757f5de356ee267e3f9f794e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Light yellow to Light orange powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralisation titration) min. 98.0 %
Melting point gt. 44.0 and le 48.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR