4'-Methyl-4-biphenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

Synonyms: (4'-Methyl-[1,1'-biphenyl]-4-yl)boronic acid

M3235-1G M3235-5G
TCM3235-1GEA 94.76 USD
TCM3235-1G TCM3235-5G
4'-Methyl-4-biphenylboronic acid (contains varying amounts of Anhydride)
4'-Methyl-4-biphenylboronic acid
Formula: C₁₃H₁₃BO₂
MW: 212.06 g/mol
Melting Pt: 138 °C
MDL Number: MFCD04038170
CAS Number: 393870-04-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a0952e1caa7ba18cb741e9e8350cfbd36a552507&vid=334e4b76d5edcb4ebb189c7ea24a59dbb6b20d50&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 110.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR