4-Methyl-3-pyridineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 4-Methylpyridine-3-boronic acid pinacol ester, 2-(4-Methyl-3-pyridyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-Methyl-3-pyridylboronic acid Pinacol ester, 4-Methyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

M3231-1G M3231-5G
TCM3231-1GEA 95.79 USD
TCM3231-1G TCM3231-5G
4-Methyl-3-pyridineboronic acid pinacol ester ≥98.0% (by GC, titration analysis)
4-Methyl-3-pyridineboronic acid pinacol ester
Formula: C₁₂H₁₈BNO₂
MDL Number: MFCD12923382
CAS Number: 1171891-31-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7ade8a1b39416d82c8982eb3374aba90713ca553&vid=6bdddd856fe1c28ae6283afc692de91c2d3f2e76&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralisation titration) min. 98.0 %
Melting point 106.0 to 110.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR