Methoxy(dimethyl)-n-octylsilane ≥95.0% (by GC)

Supplier: TCI America
Warning

Synonyms: Methoxydimethyl(octyl)silane

M3217-25ML M3217-5ML
TCM3217-25MLEA 196.73 USD
TCM3217-25ML TCM3217-5ML
Methoxy(dimethyl)-n-octylsilane ≥95.0% (by GC)
Methoxy(dimethyl)-n-octylsilane
Formula: C₁₁H₂₆OSi
MW: 202.41 g/mol
Flash Pt: 83 °C
MDL Number: MFCD00043322
CAS Number: 93804-29-6
EINECS: 298-404-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=00a1e632297a95758791ae8d0f07d9ede10c0c54&vid=796198d16d7e43d514b774b0108dc4c7fbf38c92&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR