2-Methyl-3-butenoic acid ≥96.0% (by GC, titration analysis)

Supplier: TCI America
Danger

Synonyms: 2-Methylbut-3-enoic acid

M3188-1G M3188-5G
TCM3188-1GEA 98.88 USD
TCM3188-1G TCM3188-5G
2-Methyl-3-butenoic acid ≥96.0% (by GC, titration analysis)
2-Methyl-3-butenoic acid
Formula: C₅H₈O₂
MW: 100.12 g/mol
Boiling Pt: 94 °C
MDL Number: MFCD00042650
CAS Number: 53774-20-2
UN: 3265
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c33ae13664ecb6849c74db917a42417cb8d03227&vid=766f732ffc8be66271b4cc9e9a5e85f5f5eafbbb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 96.0 %
Purity (Neutralisation titration) min. 96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR