1-Methyl-4-nitroimidazole ≥98.0% (by GC)

Supplier: TCI America
Warning

M3171-1G M3171-5G
TCM3171-1GEA 60.77 USD
TCM3171-1G TCM3171-5G
1-Methyl-4-nitroimidazole ≥98.0% (by GC)
1-Methyl-4-nitroimidazole
Formula: C₄H₅N₃O₂
MDL Number: MFCD00159683
CAS Number: 3034-41-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=09cc873a613f16ca6a3386fc927455c0f9622be3&vid=13907a27ca5f6a07deb5359667fba309100b24ca&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 134.0 to 138.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR